Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Head, Neck and Shoulder Cover for M-100 and M-300 Series PAPR

Catalog No. 19039544 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo™ M-100 and M-300 Series
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-039-544 Versaflo™ M-100 and M-300 Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-039-544 Supplier 3M™ Supplier No. 7000030015

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

For use with M-100 and M-300 Series PAPR

  • Material: Single Layer Polypropylene
  • 3M Catalog# M-976

TRUSTED_SUSTAINABILITY

Specifications

Color White
Disposable Reusable
Material Polypropylene
Product Type Head, Neck, Shoulder Cover
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo™ M-100 and M-300 Series
Model M-976
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.