Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ M-100 Series PAPR Faceshield Accessory, Head Insert

Catalog No. 19039513 Shop All 3M Products
Change view
Click to view available options
Faceshield inserts
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-039-513 Faceshield inserts
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-039-513 Supplier 3M™ Supplier No. 7100004874

missing translation for 'validMainframeMsg'

Inserts can be snapped into a Versaflo™ M-100 Series faceshields to create a bumpcap.

Helps protect the wearer's head from overspray, dust, dirt, or debris

  • A pair of inserts (left and right) are included
  • 3M Catalog# M-170

TRUSTED_SUSTAINABILITY

Specifications

Color White
Disposable Reusable
Product Type Head Insert for Faceshield
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo M-100 Series PAPR Faceshields
Model M-170
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.