Learn More
3M™ Versaflo™ TR-838 Battery Attachment Tool
Catalog No. 17000731 Shop All 3M Products
Change view
Click to view available options
Product Type:
Battery Pack
Catalog No. Product Type
17-000-731 Battery Pack
1 options
This item is not returnable. View return policy
Catalog No. 17-000-731 Supplier 3M™ Supplier No. 7100150921
Add to Cart
Add to Cart
This item is not returnable. View return policy
Versaflo™ TR-838 Battery Attachment Tool is used to mechanically attach TR-830 batteries to the TR-802N Intrinsically Safe PAPR blower motor as required by the most current UL standard (8th edition UL 913) for Intrinsically Safe PAPRs.
  • Enables attachment of TR-830 batteries to TR-802N PAPR blower motors
  • 3M Catalog# TR-838
Certifications
NIOSH Approved
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH Approved
Product Type Battery Pack
For Use With (Equipment) Versaflo™ TR-800 Series PAPR Batteries
Model TR-838