Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Breathing Tube Radiant Heat Cover, BT-927N

Catalog No. 17000743 Shop All 3M Products
Catalog No. 17-000-743 Supplier 3M™ Supplier No. 7100154106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Radiant heat resistant, high durability breathing tube cover helps extend the life of the tube.

Features:

  • Radiant heat resistant
  • Modular
  • Breathing tube cover
  • Protects against radiant heat
  • 3M Catalog# BT-927N

Certifications

NIOSH Approved

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
For Use With (Application) Assembly
Model BT-927N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.