Learn More
3M™ Versaflo™ Length-Adjusting Breathing Tube
Catalog No. 19039502 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo™ TR-300 Series PAPR
Catalog No. For Use With (Equipment)
19-039-502 Versaflo™ TR-300 Series PAPR
1 options
This item is not returnable. View return policy
Catalog No. 19-039-502 Supplier 3M™ Supplier No. 7100009685
Add to Cart
Add to Cart
This item is not returnable. View return policy
Features a flexible helix that adjusts to the appropriate length for the individual wearer.
  • QRS (Quick Release Swivel) breathing tubes allow fast and easy connection
  • Meets higher respiratory performance requirements with 3M™ headtops
  • Special flame-resistant sleeve lengthens life of tube and protects it from flying sparks
  • Disposable breathing tube cover is available to help cleaning and decontamination operations
  • 3M Catalog# BT-30
TRUSTED_SUSTAINABILITY
Specifications
Product Type Breathing Tube
For Use With (Application) Assembly, facility maintenance, grinding, machine operation, painting, sanding
For Use With (Equipment) Versaflo™ TR-300 Series PAPR
Model BT-30