Learn More
3M™ Versaflo™ Heavy Duty Neoprene Rubber Breathing Tube
Catalog No. 19039503 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo™ TR-300 Series PAPR
Catalog No. For Use With (Equipment)
19-039-503 Versaflo™ TR-300 Series PAPR
1 options
Catalog No. 19-039-503 Supplier 3M™ Supplier No. 7100105845
Add to Cart
Add to Cart
Breathing tube made of heat-resistant neoprene rubber material for rugged applications.
  • High durability air breathing tube that is easy to wipe down
  • Quick release swivel enables one-handed connection and release.
  • Compatible with all 3M™ Versaflo™ Headtops and Air Delivery Units
  • Size: 840 mm
  • 3M Catalog# BT-40
TRUSTED_SUSTAINABILITY
Specifications
Material Neoprene
Product Type Breathing Tube
For Use With (Application) Assembly, facility maintenance, grinding, machine operation, painting, sanding
For Use With (Equipment) Versaflo™ TR-300 Series PAPR
Model BT-40