Learn More
3M™ Versaflo™ H-Series PAPR Accessory, Breathing Tube Adapter
Catalog No. 19039505 Shop All 3M Products
Change view
Click to view available options
For 3M Versaflo breathing tubes
Catalog No.
19-039-505 For 3M Versaflo breathing tubes
1 options
Catalog No. 19-039-505 Supplier 3M™ Supplier No. 7000127718
H-120 adapter allows H-Series Hoods to connnect to Versaflo™ breathing tubes.
  • Consists of a hood connector and a clip.
  • 3M Catalog# H-120
TRUSTED_SUSTAINABILITY
Specifications
Product Type Breathing Tube Adapter
For Use With (Application) Versaflo™ breathing tubes
For Use With (Equipment) H-Series Headgear, Versaflo™ Breathing Tube
Includes Hood connector and clip
Model H-120