Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ M-100 Series PAPR Accessory, Forehead Seal

Catalog No. 19039512 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo M-100 Series PAPR Systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-039-512 Versaflo M-100 Series PAPR Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-039-512 Supplier 3M™ Supplier No. 7000127602
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Forehead seal is a replacement part for Versaflo™ M-100 Series faceshields.

  • 3M Catalog# M-154

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Disposable Reusable
Product Type Forehead Seal
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo M-100 Series PAPR Systems
Model M-154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.