Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ M-300 Series PAPR Accessory, Airflow Deflector

Catalog No. 19039517 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo M-300 Series Hardhats and M-400 Series Helmets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-039-517 Versaflo M-300 Series Hardhats and M-400 Series Helmets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-039-517 Supplier 3M™ Supplier No. 7000127678
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Airflow deflector is a replacement part for Versaflo™ M-300 Series hardhats and M-400 Series helmets.

  • 3M Catalog# M-316

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Disposable Reusable
Product Type Airflow Deflector
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo M-300 Series Hardhats and M-400 Series Helmets
Model M-316
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.