Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ M-400 Series PAPR Inner Collar

Catalog No. 19039525 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-400 Helmet Shrouds
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-039-525 M-400 Helmet Shrouds
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-039-525 Supplier 3M™ Supplier No. 7000127683

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inner collar zips into all Versaflo™ M-Series shrouds and provides a comfortable respiratory seal for M-400 Series helmets.

  • The M-444 Inner Collar is a required part of a M-400 system.
  • 3M Catalog# M-444

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Disposable Reusable
Product Type Inner Collar
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) M-400 Helmet Shrouds
Model M-444
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.