Learn More
3M™ Versaflo™ Gasket for M-Series PAPR Visor
Catalog No. 19039532 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-Series PAPR Systems
Catalog No. For Use With (Equipment)
19-039-532 M-Series PAPR Systems
1 options
Catalog No. 19-039-532 Supplier 3M™ Supplier No. 7000127688
  • $12.00
    $10.65 / Each
    Save $1.35
    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $59.75
    $53.65 / Case of 5 EA
    Save $6.10
    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For use with M-Series PAPR visors.
  • For use with M-Series PAPR visors
  • 3M Catalog# M-921
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Material Polycarbonate
Product Type Visor Gasket
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) M-Series PAPR Systems
Model M-921