Learn More
3M™ Versaflo™ Over-Visor Frame, M-961N
Catalog No. 17000741 Shop All 3M Products
Change view
Click to view available options
3M™ Versaflo™ Over-visor Frame, M-961N
Catalog No.
17-000-741 3M™ Versaflo™ Over-visor Frame, M-961N
1 options
Catalog No. 17-000-741 Supplier 3M™ Supplier No. 7100154127
Add to Cart
Add to Cart
Carrier frame to fit the Versaflo™ over-visor M-967N to M-Series Helmets, M-307 and M-407.
Features:
  • Designed for 3M™ Versaflo™ M-307 and M-407 helmets
  • Modular
  • Low profile
  • Over-visor
  • 3M Catalog# M-961N
Certifications
NIOSH Approved
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH Approved
Disposable Reusable
For Use With (Equipment) M-Series Helmets, M-307 and M-407
Model M-961 N