Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ PAPR Unit TR-302N+

Catalog No. 17000719 Shop All 3M Products
Change view
Click to view available options
3M™ Versaflo™ PAPR Unit TR-302N+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-000-719 3M™ Versaflo™ PAPR Unit TR-302N+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-000-719 Supplier 3M™ Supplier No. 7100153809
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Versaflo™ TR-300N+ PAPR combines performance, protection and functionality with a slim, lightweight balanced design for comfort.

Features:

  • Two selectable flow rate options
  • Easy-to-use and versatile PAPR
  • Offers more options and comfort to the user
  • For particulate environments
  • 3M Catalog# TR-302N+

Certifications

NIOSH Approved

TRUSTED_SUSTAINABILITY

Specifications

Battery Life 6 hours
Battery Type Lithium-ion
Certifications/Compliance NIOSH Approved
Color Beige
Product Type PAPR Assembly, No Headtop
Flow Rate 6.5 CFM and 7.2 CFM
For Use With (Application) Powder Handling
Model TR-302N+
Mounting Type Belt-mounted

Does not include a filter, belt, battery, or charger.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.