Learn More
3M™ Versaflo™ TR-800-PSK Painters PAPR Assembly Painters Kit
Catalog No. 17000735 Shop All 3M Products
Change view
Click to view available options
3M™ Versaflo™ Powered Air Purifying Respirator Assembly Painters Kit TR-800-PSK/94248(AAD)
Catalog No.
17-000-735 3M™ Versaflo™ Powered Air Purifying Respirator Assembly Painters Kit TR-800-PSK/94248(AAD)
1 options
This item is not returnable. View return policy
Catalog No. 17-000-735 Supplier 3M™ Supplier No. 7100150922
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Faceshield Assembly, Breathing Tube, OV/HE Cartridge, Battery Charger, Tube Cover, IS PAPR Unit, Battery Attachment Tool, Easy Clean Belt, Filter Cover, Airflow Indicator, Storage and Cleaning Kit.
3M™ Versaflo™ TR-800-PSK is a complete Powered Air Purifying Respirator with an Intrinsically Safe motor/blower that includes everything required to get started for use in paint spray operations.
Features:
  • Top of the line IS protection - The TR-800 is a PAPR blower rated at Division I for Classes I,II, and III under the most current UL standard (UL 60079, 6th Edition, 2013)
  • More options for more tasks - Choose from particulate filters or gas and vapor combination cartridges for protection in a variety of environments.
  • Work at your own speed - Multiple airflow rates put you in control of your comfort.
  • When clean is critical - Easy-to-clean belt and disposable breathing tube covers save time and effort, paving the way for more productivity.
  • 3M Catalog# TR-800-PSK
Certifications
NIOSH Approved
Compliance
UL-certified Intrinsically Safe for Division 1: Class I, II, III; Division 1 (includes Division 2) Groups C, D, E, F, G; T4, under the most current standard (UL 60079, 6th Edition, 2013).
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 6 to 10 hours
Battery Type Lithium-ion
Certifications/Compliance UL-Certified Intrinsically Safe for Division 1: Class I, II, III, Division 1 (includes Division 2) Groups C, D, E, F, G, T4 (UL 60079, 6th Edition, 2013)
Filter Type HEPA
Product Type PAPR Assembly Painters Kit
For Use With (Application) Painting; Powder Coating; Spray Painting
Model TR-800-PSK
Mounting Type Belt-mounted