Learn More
3M™ Versaflo™ Powered Air Purifying Respirator Easy Clean Assembly TR-814N/94247(AAD)
Catalog No. 17000734 Shop All 3M Products
Change view
Click to view available options
3M™ Versaflo™ Powered Air Purifying Respirator Easy Clean Assembly TR-814N/94247(AAD)
Catalog No.
17-000-734 3M™ Versaflo™ Powered Air Purifying Respirator Easy Clean Assembly TR-814N/94247(AAD)
1 options
This item is not returnable. View return policy
Catalog No. 17-000-734 Supplier 3M™ Supplier No. 7100150923
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Intrinsically Safe motor/blower (TR-802N), Intrinsically Safe Battery (TR-830N), battery attachment tool (TR-838), Airflow Indicator (TR-971), Easy Clean Belt (TR-627) and Length Adjusting Breathing Tube (BT-30).
The 3M™ Versaflo™ Powered Air Purifying Respirator, TR-800 is a PAPR blower rated at Division I for Classes I,II, and III under the most current UL standard (UL 60079, sixth Edition, 2013).
Features:
  • Top of the line IS protection - The TR-800 is a PAPR blower rated at Division I for Classes I,II, and III under the most current UL standard (UL 60079, 6th Edition, 2013)
  • More options for more tasks - Choose from particulate filters or gas and vapor combination cartridges for protection in a variety of environments.
  • Work at your own speed - Multiple airflow rates put you in control of your comfort.
  • When clean is critical - Easy-to-clean belt and optional disposable breathing tube covers and storage and cleaning kit save time and effort, paving the way for more productivity.
  • 3M Catalog# TR-814N
Certifications
NIOSH Approved
TRUSTED_SUSTAINABILITY
Specifications
Battery Type Lithium-ion
Certifications/Compliance NIOSH Approved
Product Type PAPR Assembly, No Headtop
For Use With (Application) Liquid Chemical Handling; Powder Handling; Powder Coating; Spray Painting; General Maintenance; Painting; Mixing; Cleaning
Includes Intrinsically Safe motor/blower (TR-802N), Intrinsically Safe Battery (TR-830N), battery attachment tool (TR-838), Airflow Indicator (TR-971), Easy Clean Belt (TR-627) and Length Adjusting Breathing Tube (BT-30).
Model TR-814N
Mounting Type Belt-mounted