Learn More
3M™ Versaflo™ TR-813N Heavy-Duty PAPR Assembly
Catalog No. 17000733 Shop All 3M Products
Change view
Click to view available options
3M™ Versaflo™ Powered Air Purifying Respirator Heavy Duty Assembly TR-813N/94246(AAD)
Catalog No.
17-000-733 3M™ Versaflo™ Powered Air Purifying Respirator Heavy Duty Assembly TR-813N/94246(AAD)
1 options
This item is not returnable. View return policy
Catalog No. 17-000-733 Supplier 3M™ Supplier No. 7100150924
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Instrinsically Safe motor/blower (TR-802N), Intrinsically Safe Battery (TR-830N), battery attachment tool (TR-838), Airflow Indicator (TR-971), High Durability Belt (TR-626) and Length Adjusting Breathing Tube (BT-30).
Features:
  • Top of the line IS protection - The TR-800 is a PAPR blower rated at Division I for Classes I,II, and III under the most current UL standard (UL 60079, 6th Edition, 2013)
  • Cover your bases - Utilize cartridges to help protect against certain gas and vapors, combined with filters for protection against particulates.
  • Feel heightened comfort - Heavy-duty belt designed with airflow channels, multiple size adjustments and belt extender options.
  • Work with ease - Intuitive, colored touchpoints plus visual and audible alarms  help make the TR-800 easy to use.
  • Keep the air flowing - Multi-speed blower functions up to 16,000 feet and battery offers long run time, less charge time and, ultimately, reduced down time.
  • 3M Catalog# TR-813N
Certifications
NIOSH Approved
Compliance
UL-certified Intrinsically Safe for Division I: IS Class I, II, III; Division I (includes Division 2) Groups C, D, E, F, G; T4 under the most current standard (UL 60079, 6th edition, 2013).
TRUSTED_SUSTAINABILITY
Specifications
Battery Type Lithium-ion
Certifications/Compliance UL-Certified Intrinsically Safe for Division 1: Class I, II, III, Division 1 (includes Division 2) Groups C, D, E, F, G, T4 (UL 60079, 6th Edition, 2013)
Product Type PAPR System for Heavy Industry
For Use With (Application) General Maintenance; Spray Painting; Cleaning; Powder Coating; Mixing; Liquid Chemical Handling; Powder Handling; Painting
Includes Instrinsically Safe motor/blower (TR-802N), Intrinsically Safe Battery (TR-830N), battery attachment tool (TR-838), Airflow Indicator (TR-971), High Durability Belt (TR-626) and Length Adjusting Breathing Tube (BT-30).
Model TR-813N
Mounting Type Belt-mounted