Learn More
3M™ Versaflo™ TR-800-HIK Heavy Industry PAPR Assembly Kit
Catalog No. 17000737 Shop All 3M Products
Change view
Click to view available options
3M™ Versaflo™ Powered Air Purifying Respirator Heavy Industry Kit TR-800-HIK
Catalog No.
17-000-737 3M™ Versaflo™ Powered Air Purifying Respirator Heavy Industry Kit TR-800-HIK
1 options
Catalog No. 17-000-737 Supplier 3M™ Supplier No. 7100150927
Add to Cart
Add to Cart
Includes
Hard Hat Assembly, IS PAPR Unit, Battery, Battery Attachment Tool, Breathing Tube, High Durability Belt, OV/AG/HE Cartridge, Filter Cover, Battery Charger Kit, Airflow Indicator, Prefilter, Spark Arrestor, Storage and Cleaning Kit.
3M™ Versaflo™ TR-800-HIK is a complete Powered Air Purifying Respirator featuring an Intrinsically Safe motor/blower and including everything required to get started
Features:
  • Top of the line IS protection - The TR-800 is a PAPR blower rated at Division I for Classes I,II, and III under the most current UL standard (UL 60079, 6th Edition, 2013)
  • Cover your bases - Utilize cartridges to help protect against certain gas and vapors, combined with filters for protection against particulates.
  • Fewer worries - Rugged, durable faceshield with premium visor helps protect against sparks and debris.
  • Feel heightened comfort - Heavy-duty belt designed with airflow channels, multiple size adjustments and belt extender options.
  • Work with ease - Intuitive, colored touchpoints plus visual and audible lowflow indicators help make the TR-800 easy to use.
  • Keep the air flowing - Multi-speed blower functions up to 16,000 feet and  battery offers long run time, less charge time and, ultimately, reduced down time.
  • Simple way to purchase a complete PAPR system for use in rugged industrial environments.
  • 3M Catalog# TR-800-HIK
Certifications
NIOSH Approved
Compliance
UL-certified Intrinsically Safe for Division 1: Class I, II, III; Division 1 (includes Division 2) Groups C, D, E, F, G; T4, under the most current standard (UL 60079, 6th Edition, 2013).
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 6 to 10 hours
Battery Type Lithium-ion
Certifications/Compliance UL-Certified Intrinsically Safe for Division 1: Class I, II, III, Division 1 (includes Division 2) Groups C, D, E, F, G, T4 (UL 60079, 6th Edition, 2013)
Filter Type HEPA
Product Type PAPR System for Heavy Industry
For Use With (Application) Powder Handling; Liquid Chemical Handling; Powder Coating; Painting; General Maintenance; Spray Painting; Cleaning; Mixing
Model TR-800-HIK
Mounting Type Belt-mounted