Learn More
3M™ Versaflo™ TR-802N Motor/Blower
Catalog No. 17000738 Shop All 3M Products
Change view
Click to view available options
Product Type:
Motor Blower
Catalog No. Product Type
17-000-738 Motor Blower
1 options
Catalog No. 17-000-738 Supplier 3M™ Supplier No. 7100150821
Add to Cart
Add to Cart
Versaflo™ Powered Air Purifying Respirator Unit TR-802N/94242(AAD) is an intrinsically safe replacement motor/blower for 3M Versaflo TR-800 series Powered Air Purifying Respirators.
Features:
  • Top of the line IS protection - The TR-800 is a PAPR blower rated at Division I for Classes I,II, and III under the most current UL standard (UL 60079, 6th Edition, 2013)
  • More options for more tasks - Choose from particulate filters or gas and vapor combination cartridges for protection in a variety of environments.
  • Work with ease - Intuitive, colored touchpoints plus visual and audible low flow indicators help make the TR-800 easy to use.
  • Work at your own speed - Multiple airflow rates put you in control of your comfort.
  • When clean is critical - Easy-to-clean belt and disposable breathing tube covers save time and effort, paving the way for more productivity.
  • Combine with 3M Versaflo™ components to build a complete system.
  • 3M Catalog# TR-802N
Certifications
NIOSH Approved
Compliance
UL-certified Intrinsically Safe (UL 60079, 6th Edition, 2013).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Motor Blower
For Use With (Application) General maintenance, cleaning, powder handling, painting, liquid chemical handling, mixing
For Use With (Equipment) Versaflo™ TR-800 PAPR Systems
Model TR-802N
Mounting Type Belt-mounted
Does not include a filter, belt, battery, or charger