Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Radiant Heat Cover

Catalog No. 17000740 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo™ M-306 and M-407 Helmet
Versaflo™ M-307 Hard Hat Assembly
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-000-740 Versaflo™ M-306 and M-407 Helmet
17-000-739 Versaflo™ M-307 Hard Hat Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-000-740 Supplier 3M™ Supplier No. 7100154130
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to be used with the 3M™ Versaflo™ helmet in high temperature environments. Provides head, neck, and shoulder protection and comfort from radiant heat.

Features:

  • Modular range allows for multiple combinations and different coverage levels
  • Compatible with 3M™ Versaflo™ Helmet
  • Modular
  • Protects against radiant heat

Certifications

NIOSH Approved

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH Approved
Color Silver
Disposable Reusable
Material Aluminized
For Use With (Application) Foundries
For Use With (Equipment) Versaflo™ M-306 and M-407 Helmet
Model M-975 N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.