Learn More
3M™ Versaflo™ TR-600/800 PAPR Accessory, Cleaning and Storage Kit TR-653 / 37373 (AAD)
Catalog No. 19268028 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
TR-600/TR-800 PAPR System
Catalog No. For Use With (Equipment)
19-268-028 TR-600/TR-800 PAPR System
1 options
Catalog No. 19-268-028 Supplier 3M™ Supplier No. 7100035841
Add to Cart
Add to Cart
Includes
1 inlet plug and 2 seals, 1 outlet plug and 2 seals and 1 battery connector cover
Protects from internal contamination during cleaning and storage.
  • Plug and seal for inlet and outlet openings of TR-600 series motor/blower
  • Cover for electrical connections of TR-630 or TR-632 battery
  • Motor/blower plugs allow motor/blower to be submerged, rinsed under running water, or machine washed
  • Plug seals to be replaced after 30 uses or whenever found to be damaged or worn
  • 3M Catalog# TR-653
Applications:
Recommended Industry: Healthcare, Pharmaceutical, Pesticide Application, Heavy Industrial, Animal processing, Laboratories, Nuclear energy, Manufacturing, Cosmetics, Pharmaceutical research and production, Recycling, General Manufacturing, Dry Foods
Certifications
NIOSH Approved
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Product Type Cleaning and Storage Plug
For Use With (Equipment) TR-600/TR-800 PAPR System
Includes 1 inlet plug and 2 seals, 1 outlet plug and 2 seals, and 1 battery connector cover
Model TR-653
Quantity 1 Ea.