Learn More
3M™ Versaflo™ TR-600/800 Series PAPR Accessory, Battery Charger Cradle TR-640
Catalog No. 19268023 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
TR-600/800 Series PAPR Systems
Catalog No. For Use With (Equipment)
19-268-023 TR-600/800 Series PAPR Systems
1 options
Catalog No. 19-268-023 Supplier 3M™ Supplier No. 7100035837
Provides easy-to-use interface between all TR-600 Series batteries and all TR-900 Series power supplies.
  • Replacement part for TR-641 and TR-644 battery charger kits
  • Holds TR-630 or TR-632 batteries
  • Can be used on TR-944 4 station power supply
  • 3M Catalog# TR-640
Applications:
Recommended Industry: Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Certifications
NIOSH Approved
TRUSTED_SUSTAINABILITY
Specifications
Product Type Single-Unit Charging Station
For Use With (Equipment) TR-600/800 Series PAPR Systems
Model TR-640
No. of Ports One
Quantity 1/Cs.