Learn More
3M™ Versaflo™ TR-600-ECK Easy Clean PAPR Assembly Kit
Catalog No. 19268006 Shop All 3M Products
Change view
Click to view available options
For Use With (Application):
Pharmaceutical manufacturing, health care, food processing, painting, and lead battery manufacturing/recycling
Catalog No. For Use With (Application)
19-268-006 Pharmaceutical manufacturing, health care, food processing, painting, and lead battery manufacturing/recycling
1 options
Catalog No. 19-268-006 Supplier 3M™ Supplier No. 7100035754
  • $3,752.00
    $3,023.65 / Case of 1
    Save $728.35
    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Complete powered air purifying respirator includes all required components to get started.
  • Complete easy-to-use PAPR
  • Hood has integrated suspension for easy use and maintenance
  • Kit Components
    • S-433 Hood assembly
    • TR-614 PAPR assembly [includes TR-602N motor/blower, TR630 standard battery, BT-30 Length Adjusting Breathing Tube, TR-627 Easy Clean Belt, TR-971 Airflow indicator] (FS 19-268-002)
    • TR-6710N (HE) Filter for particulates (FS 19-268-009)
    • TR-6700FC Filter cover (FS 19-268-033)
    • TR- 641 Single station battery charger (FS 19-268-024)
    • BT-922 Disposable breathing tube cover
    • Applications:
      Pharmaceutical manufacturing, health care, food processing, painting, and lead battery manufacturing/recycling
    • 3M Catalog# TR-600-ECK
Compliance
NIOSH approved.
TRUSTED_SUSTAINABILITY
Specifications
Product Type PAPR System with Hood
For Use With (Application) Pharmaceutical manufacturing, health care, food processing, painting, and lead battery manufacturing/recycling
Includes Hood assembly, PAPR Assembly, filter for particulates, filter cover, single station battery charger, disposable breathing tube cover
Model TR-600-ECK