Learn More
3M™ Versaflo™ TR-600-HIK Heavy Industry PAPR Kit
Catalog No. 19268005 Shop All 3M Products
Change view
Click to view available options
For Use With (Application):
Industrial, demolition, grinding, metal finishing and casting operations
Catalog No. For Use With (Application)
19-268-005 Industrial, demolition, grinding, metal finishing and casting operations
1 options
Catalog No. 19-268-005 Supplier 3M™ Supplier No. 7100035755
Add to Cart
Add to Cart
Complete powered air purifying respirator includes all required components to get started.
  • Easy to use
  • Provides comfortable protection
  • Kit Components
    • M-307 Headtop [hard hat assembly] (FS Cat. No. 19-039-516 )
    • TR-616 PAPR assembly [includes TR-602N motor/blower, BT-30 Length Adjusting Breathing Tube, TR-626 High Durability Belt] (FS 19-268-003)
    • TR-632 High capacity battery (FS 19-268-022)
    • TR-971 Airflow indicator (FS 19-268-035)
    • TR-6530N (HE/OV/AG/HF) Filter/Cartridge (FS 19-268-011)
    • TR-6300FC Filter cover (FS 19-268-021)
    • TR- 641 Single station battery charger (FS 19-268-024)
    • BT-926 High temperature breathing tube cover
    • Applications:
      Industrial, demolition, grinding, metal finishing and casting operations
    • 3M Catalog# TR-600-HIK
TRUSTED_SUSTAINABILITY
Specifications
Product Type PAPR System for Heavy Industry
For Use With (Application) Industrial, demolition, grinding, metal finishing and casting operations
Includes Headtop, PAPR assembly (motor/blower, breathing tube, high durability belt, high capacity battery, airflow indicator), filter/cartridge, filter cover, single station battery charger, high temperature breathing tube cover
Model TR-600-HIK