Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Back Pack for TR-600/800 PAPR

Catalog No. 19268030 Shop All 3M Products
Change view
Click to view available options
TR-655 Back Pack Adaptor; For TR-600 series PAPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-268-030 TR-655 Back Pack Adaptor; For TR-600 series PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-268-030 Supplier 3M™ Supplier No. 7000052901

missing translation for 'validMainframeMsg'

Backpack allows wearer to carry TR-600 Series PAPR on the back rather than waist-mounted belt. The TR-600 PAPR system is securely attached via the TR-655 Back Pack Adapter (19-072-000).

  • Allows user to wear the TR-600 series PAPR on their back; workers may find the PAPR more comfortable or convenient to wear when mounted on the backpack. 
  • This configuration is particularly suitable when a worker already wears other equipment around their waist
  • Multi-layer construction secures TR-600 in place
  • For Powder Handling, Sawing, Grinding, Powder Coating, Cutting, Drilling, Sweeping, Infection Control, Mixing, General Maintenance, Cleaning
  • Color: Black
  • The TR-655 back pack adapter (19-268-030), enables the TR-600 series PAPR to be worn on the BPK-01 backpack
  • 3M Catalog# BPK-01

Certifications

NIOSH Approved

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH Approved
Color Black
Disposable Reusable
Product Type Backpack
For Use With (Application) Powder Handling, Sawing, Grinding, Powder Coating, Cutting, Drilling, Sweeping, Infection Control, Mixing, General Maintenance, Cleaning
For Use With (Equipment) Versaflo™ TR-600 Series PAPR System
Model BPK-01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.