Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ TR-600 Series PAPR Accessory, Spark Arrestor

Catalog No. 19268032 Shop All 3M Products
Change view
Click to view available options
Quantity:
2/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-268-032 2/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-268-032 Supplier 3M™ Supplier No. 7100035845
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Versaflo™ TR-600 Series PAPR.

  • TR-662 spark arrestor is used with all TR-600 series filters and cartridges
  • Protects TR-6000 series filters and cartridges from potential damage due to sparks
  • Acts as pre-filter by capturing larger particles that would load main filter
  • Flexible application as TR-662 may be used with TR-6600 prefilter
  • Reduces operating costs by extending life of TR-6000 series filters and cartridges
  • Color: Silver
  • 3M Catalog# TR-662

Applications:

Healthcare

Certifications

NIOSH Approved

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Disposable Reusable
Product Type Spark Arrestor
For Use With (Application) Healthcare
For Use With (Equipment) Versaflo™ TR-600 Series PAPR System
Model TR-662
Quantity 2/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.