Learn More
3M™ Versaflo™ TR-602N Motor Blower
Catalog No. 19268000 Shop All 3M Products
Change view
Click to view available options
Product Type:
Motor Blower
Catalog No. Product Type
19-268-000 Motor Blower
1 options
Catalog No. 19-268-000 Supplier 3M™ Supplier No. 7100035756
Add to Cart
Add to Cart
Replacement motor/blower for 3M Versaflo TR-600 Series Powered Air Purifying Respirators
  • Versatile for a wide range of applications
  • Comfortable ergonomic design
  • User selectable airflows
  • Easy to use and maintain
  • 3M Catalog# TR-602N
Order Info
Does not include a filter, airflow indicator, belt, battery, or charger.
Compliance
NIOSH approved.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Motor Blower