Learn More
3M™ Versaflo™ TR-613N PAPR Assembly
Catalog No. 19268001 Shop All 3M Products
Change view
Click to view available options
For Use With (Application):
Grinding and metal finishing applications.
Catalog No. For Use With (Application)
19-268-001 Grinding and metal finishing applications.
1 options
Catalog No. 19-268-001 Supplier 3M™ Supplier No. 7100035757
Add to Cart
Add to Cart
Powered air purifying respirator provides durable protection even in harsh environments.
The TR-613N assembly may be combined with 3M Versaflo branded headtops and filters or cartridges to create a complete, compact, light weight, easy-to-use PAPR for protection against certain particulates, vapors and gases. Single and four bay chargers are available to charge the battery included in the TR-613N assembly. The TR-613N assembly may be the preferred choice for working in grinding and metal finishing applications.
  • Assembly configured of commonly used components
  • Easy to use
  • Lightweight, durable belt
  • Assembly Components
    • Motor/blower (FS Cat. No. 19-268-000)
    • Standard capacity rechargeable lithium ion battery (FS Cat. No. 19-268-020)
    • High durability belt (FS Cat. No. 19-268-016)
    • Airflow indicator (FS Cat. No. 19-268-035)
    • 3M Catalog# TR-613N
Order Info
Headtops, filters, cartridges and battery charger sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Battery Type Lithium-ion
Product Type PAPR Assembly, No Headtop
For Use With (Application) Grinding and metal finishing applications.
Includes Motor/blower, standard battery, length adjusting breathing tube, high durability belt, airflow indicator
Model TR-613N