Learn More
3M™ Versaflo™ TR-617N/37342(AAD) PAPR Assembly
Catalog No. 19268004 Shop All 3M Products
Change view
Click to view available options
For Use With (Application):
Pharmaceutical or food manufacturing
Catalog No. For Use With (Application)
19-268-004 Pharmaceutical or food manufacturing
1 options
Catalog No. 19-268-004 Supplier 3M™ Supplier No. 7100035830
Add to Cart
Add to Cart
Powered air purifying respirator provides durable protection even in harsh environments.
The TR-617N assembly may be combined with 3M Versaflo branded headtops and filters or cartridges to create a complete, compact, light weight, easy-to-use PAPR for protection against certain particulates, vapors and gases. Single and four bay chargers are available to charge the battery included in the TR-616N assembly. The TR-617N assembly may be the prefered choice for working in rugged environments with airborne hazards that include particulates and gases or vapors such as pharmaceutical or food manufacturing.
  • Assembly configured of commonly used components
  • Easy to use and clean
  • Long run times
  • Assembly Components
    • Motor/blower (FS Cat. No. 19-268-000)
    • Length adjusting breathing tube (FS Cat. No. 19-039-502 )
    • Easy clean belt (FS Cat. No. 19-268-018)
    • High capacity rechargeable lithium ion battery (FS Cat. No. 19-268-022)
    • Airflow indicator (FS Cat. No. 19-268-035)
    • 3M Catalog# TR-617N
Order Info
Headtops, filters, cartridges and battery charger sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Battery Type High Capacity Lithium Ion
Product Type PAPR Assembly, No Headtop
For Use With (Application) Pharmaceutical or food manufacturing
Includes Motor/blower, high capacity battery, length adjusting breathing tube, high durability belt, airflow indicator
Model TR-617N