Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ TR-6590N Multi-Gas/HE Cartridge

Catalog No. 17000729 Shop All 3M Products
Change view
Click to view available options
3M™ Versaflo™ TR-6590N Multi-Gas/HE Cartridge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-000-729 3M™ Versaflo™ TR-6590N Multi-Gas/HE Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-000-729 Supplier 3M™ Supplier No. 7100035844
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Cartridge

3M™ Versaflo™ TR-6590N is a filter is a combination High Efficiency (HE) particulate filter and Multi-Gas cartridge (MG) for use with TR-600 series PAPRs

Features:

  • High efficiency particulate filter
  • Captures a broad range of certain gases and vapors, including mercury
  • Compatible with the 3M™ Versaflo™ TR-600 PAPR system
  • First multi-gas PAPR cartridge with NIOSH approval for use against mercury
  • 3M Catalog# TR-6590N

Certifications

NIOSH Approved

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Combination Cartridge/Filter
Size Standard
For Use With (Application) Liquid Chemical Handling
For Use With (Equipment) TR-600 Powered Air Purifying Respirator Systems & Versaflo™ AP-600 Portable Panels
Includes Cartridge
Model TR-6590N
Mounting Type Belt-mounted
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.