Learn More
3M™ Versaflo™ TR-Series PAPR Accessory, Spark Arrestor
Catalog No. 19039570 Shop All 3M Products
Change view
Click to view available options
Quantity:
2/Cs.
Catalog No. Quantity
19-039-570 2/Cs.
1 options
Catalog No. 19-039-570 Supplier 3M™ Supplier No. 7000052905
Add to Cart
Add to Cart
When installed inside the filter cover of the TR-300 PAPR, the TR-362 spark arrestor helps protect the HE filter and PAPR unit from damage due to sparks or other hot materials.
  • 3M Catalog# TR-362
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Product Type Spark Arrestor
For Use With (Application) Protects The he Filter and Powered Air Purifying Respirator Unit From Damage Due to Sparks or Other Hot Materials
For Use With (Equipment) Versaflo TR-Series PAPR System
Model TR-362
Quantity 2/Cs.