Learn More
Alkali Scientific 3M Vetbond Tissue Adhesive, 3ml Bottles w/MSDS (2 Bottles)
SDP
Catalog No. 5000662704
Encompass_Preferred
Creates a strong bond, polymerizes in seconds. Blue dye for easy to see drop application. Full MSDS included to assist receipt & insure safe handling. Made in the USA. 3M Vetbond quickly and conveniently closes minor wounds, often eliminating the need for sutures and/or bandages. It polymerizes in seconds after contact with tissue and body fluids and binds wound edges together so natural healing can occur. INGREDIENTS: N butyl cyanoacrylate 99.99%, Blue Dye.
Catalog No. 50-006-62704 Supplier Alkali Scientific Supplier No. TA3M
Creates a strong bond, polymerizes in seconds. Blue dye for easy to see drop application. Full MSDS included to assist receipt & insure safe handling. Made in the USA. 3M Vetbond quickly and conveniently closes minor wounds, often eliminating the need for sutures and/or bandages. It polymerizes in seconds after contact with tissue and body fluids and binds wound edges together so natural healing can occur. INGREDIENTS: N butyl cyanoacrylate 99.99%, Blue Dye.