Learn More
Hanna Pharmaceutical Supply Co., Inc. VETBOND TISSUE ADHESIVE
Catalog No. NC0304169
Encompass_Preferred
Reduces tissue trauma and need for analgesia in minor wound repairPolymerizes in seconds after contact with tissue and body fluids and binds wound edges together so natural healing can occurExcellent for quick, minor wound repairsActs as a physical barrier to keep foreign matter from entering woundSelf-sealing proprietary pin cap keeps the nozzle clear while allowing unlimited opening and resealingCat declaws Minor lacerationsAbrasionsSurgical incisionsOral surgeries Dew claw removal 3ML BOTTLE includes 4 yellow applicator tips for ease of dispensing
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC0304169 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. MMM1469SB
Reduces tissue trauma and need for analgesia in minor wound repairPolymerizes in seconds after contact with tissue and body fluids and binds wound edges together so natural healing can occurExcellent for quick, minor wound repairsActs as a physical barrier to keep foreign matter from entering woundSelf-sealing proprietary pin cap keeps the nozzle clear while allowing unlimited opening and resealingCat declaws Minor lacerationsAbrasionsSurgical incisionsOral surgeries Dew claw removal 3ML BOTTLE includes 4 yellow applicator tips for ease of dispensing
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.