Learn More
MSC 3M VHB TAPE
Catalog No. 5000796217
Encompass
VHB Tape 4950 is a double-sided tape with a general-purpose acrylic adhesive and a firm, foam core, 45 mil (1.1 mm) thick. It provides immediate handling strength, replacing mechanical fasteners, and ensures secure bonding, water protection, and long-term durability.
  • All-acrylic bonding method for long-term durability
  • Smooth, seamless alternative to mechanical fasteners
  • High shear strength and reliable surface adhesion
  • Excellent indoor and outdoor durability, up to 200 °F (93 °C)
  • Closed-cell acrylic chemistry for resistance to extreme environments
  • Vibration and fatigue resistant viscoelastic foam
Catalog No. 50-007-96217 Supplier MSC Supplier No. 02817088
VHB Tape 4950 is a double-sided tape with a general-purpose acrylic adhesive and a firm, foam core, 45 mil (1.1 mm) thick. It provides immediate handling strength, replacing mechanical fasteners, and ensures secure bonding, water protection, and long-term durability.
  • All-acrylic bonding method for long-term durability
  • Smooth, seamless alternative to mechanical fasteners
  • High shear strength and reliable surface adhesion
  • Excellent indoor and outdoor durability, up to 200 °F (93 °C)
  • Closed-cell acrylic chemistry for resistance to extreme environments
  • Vibration and fatigue resistant viscoelastic foam