Learn More
MSC 3M VINYL MARKING TAPE
Catalog No. 5000792147
Encompass
The Vinyl Tape 471 is a conformable tape with a vinyl backing and rubber adhesive, engineered for applications such as floor marking, color coding, and temporary surface protection. It provides excellent adhesion to many surfaces and allows for one-piece, clean removal.
  • Unique stretch properties conform to uneven surfaces without lifting or retracting.
  • Rubber adhesive offers excellent holding strength and quick stick on numerous substrates.
  • Good abrasion and solvent resistance ensure a long service life and color retention.
  • One-piece, clean removal from many surfaces reduces clean-up and labor costs.
  • Low leachable halogens and sulfur make it suitable for corrosion-sensitive applications.
Catalog No. 50-007-92147 Supplier MSC Supplier No. 92723477
The Vinyl Tape 471 is a conformable tape with a vinyl backing and rubber adhesive, engineered for applications such as floor marking, color coding, and temporary surface protection. It provides excellent adhesion to many surfaces and allows for one-piece, clean removal.
  • Unique stretch properties conform to uneven surfaces without lifting or retracting.
  • Rubber adhesive offers excellent holding strength and quick stick on numerous substrates.
  • Good abrasion and solvent resistance ensure a long service life and color retention.
  • One-piece, clean removal from many surfaces reduces clean-up and labor costs.
  • Low leachable halogens and sulfur make it suitable for corrosion-sensitive applications.