Learn More
MSC 3M VINYL TAPE
Catalog No. 5000802997
Encompass
3M Vinyl Tape 471 is a versatile, conformable vinyl tape with a rubber adhesive, ideal for long-lasting floor marking, paint line creation, surface protection, and reliable sealing. It conforms well to uneven surfaces without lifting and offers excellent adhesion with clean removal.
  • Vivid colors for marking and color-coding
  • Unique stretch allows conformity to uneven surfaces
  • One-piece, clean removal minimizes cleanup
  • Resistant to abrasion and solvents for long service life
  • Rubber adhesive ensures strong holding on most surfaces
  • Provides robust long-term or short-term seals
  • Low leachable halogens and sulfur for corrosion-sensitive applications
  • Color maintained despite abrasion and wear
Catalog No. 50-008-02997 Supplier MSC Supplier No. 03819257
3M Vinyl Tape 471 is a versatile, conformable vinyl tape with a rubber adhesive, ideal for long-lasting floor marking, paint line creation, surface protection, and reliable sealing. It conforms well to uneven surfaces without lifting and offers excellent adhesion with clean removal.
  • Vivid colors for marking and color-coding
  • Unique stretch allows conformity to uneven surfaces
  • One-piece, clean removal minimizes cleanup
  • Resistant to abrasion and solvents for long service life
  • Rubber adhesive ensures strong holding on most surfaces
  • Provides robust long-term or short-term seals
  • Low leachable halogens and sulfur for corrosion-sensitive applications
  • Color maintained despite abrasion and wear