Learn More
3M™ Virtua™ Reader Protective Eyewear
Catalog No. 17827303 Shop All 3M Products
Change view
Click to view available options
Magnification Power:
+2.00
Catalog No. Magnification Power
17-827-303 +2.00
1 options
This item is not returnable. View return policy
Catalog No. 17-827-303 Supplier 3M™ Supplier No. 7000127512
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for workers who have difficulty reading small print or are engaged in detail work. Unisex Virtua reader protective eyewear features magnifying diopters, lightweight lenses with contoured tip temples and a high wrap for additional coverage.
The Virtua reader protective eyewear line merges outstanding unisex style, protection and affordability, all while offering wearers an open, unobstructed viewing range. Virtua protective eyewear is a valuable choice in vision safety.
  • Clear anti-fog lens
  • Clear temple
  • Polycarbonate lens absorbs 99.9% UV
  • Bifocal readers with +2.0 magnification diopters
  • Meets the High Impact Requirements of ANSI Z87.1-2003
  • 20 safety glasses per case
  • 3M Catalog# 11514-00000-20
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010
Lens Tint Clear
Product Type Reader Protective Eyewear
Magnification Power +2.00
No. per Case 20/Cs.
Model 11514-00000-20
Lens Material Polycarbonate
UV Protection UV400