Learn More
3M™ Virtua™ Readers Eyewear
Catalog No. 19810003 Shop All 3M Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
+2.50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-810-003 +1.50
19-810-005 +2.50
19-810-004 +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-810-003 Supplier 3M™ Supplier No. 7000127511
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
For workers who have difficulty reading small print or are engaged in fine detail work
  • Bifocal readers with three magnification diopters
  • Polycarbonate lens absorbs 99.9% UV
  • Unisex styling with lightweight design weighs less than one ounce
  • High wrap for additional coverage
  • Contoured temples for comfortable fit
  • 3M Catalog# 11513-00000-20 , 3M Catalog# 11514-00000-20 , 3M Catalog# 11515-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Virtual Reader Eyewear
For Use With (Application) For workers who have difficulty reading small print or are engaged in fine detail work
Magnification Power +1.50
Model 11513-00000-20
Lens Material Polycarbonate
UV Protection UV400