Learn More
Enterprise Technology Solutions 3M Virtua Safety Glass, Blue
SDP
Catalog No. 5000828477
Encompass_Preferred
Protective eyewear features a comfortable seal that helps limit eye exposure to nuisance dust while providing additional cushioning. The anti-fog lens coating and removable foam-lined gasket with airflow vents make this protective eyewear ideal for humid environments. Foam gasket helps limit eye exposure to nuisance dust while providing additional cushioning. Polycarbonate lenses absorb 99.9 percent ultraviolent light. The comfortable, lightweight frame features the Cord Control System (CCS). CCS is designed to keep a corded earplug attached, untangled, and ready for use. Earplug is sold separately. Eyewear meets the requirements of CSA Z94.3-07 and is Impact Rated to ANSI Z87.1-2010.
Catalog No. 50-008-28477 Supplier Enterprise Technology Solutions Supplier No. MMM118720000020
Protective eyewear features a comfortable seal that helps limit eye exposure to nuisance dust while providing additional cushioning. The anti-fog lens coating and removable foam-lined gasket with airflow vents make this protective eyewear ideal for humid environments. Foam gasket helps limit eye exposure to nuisance dust while providing additional cushioning. Polycarbonate lenses absorb 99.9 percent ultraviolent light. The comfortable, lightweight frame features the Cord Control System (CCS). CCS is designed to keep a corded earplug attached, untangled, and ready for use. Earplug is sold separately. Eyewear meets the requirements of CSA Z94.3-07 and is Impact Rated to ANSI Z87.1-2010.