Learn More
3M™ Virtua™ Sport CCS Eyewear
Catalog No. 17827313 Shop All 3M Products
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 17-827-313 Supplier 3M™ Supplier No. 7010384246
Add to Cart
Add to Cart
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Corded Earplug Control System keeps a corded earplug attached, untangled and ready for use
  • Polycarbonate lenses absorb 99.9% UV
  • Compatible with a variety of vinyl-corded earplugs
  • 3M Catalog# 11796-00000-20 , 3M Catalog# 11797-00000-20 , 3M Catalog# 11798-00000-20
Compliance
Meets ANSI™ Z87. 1-2010 standard.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z87. 1-2010 standard.
Lens Tint Gray
No. per Case 20/Cs.
Model 11798-00000-20
Lens Material Polycarbonate