Learn More
3M™ Virtua™ Sport Eyewear
Catalog No. 19810009 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Hardcoat
Lens Tint:
Clear
Gray
Light Amber
Light Blue
Mirror
Frame Color:
Amber
Blue
Clear
Gray
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Frame Color
19-810-009 Hardcoat Light Blue Blue
19-810-008 Anti-fog Light Blue Blue
19-810-010 - Light Amber Amber
19-810-007 - Gray Gray
19-130-3912 - Clear Clear
19-130-3913 Anti-fog Gray Gray
19-130-3911 - Clear Clear
19-130-3915 - Mirror Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-810-009 Supplier 3M™ Supplier No. 7000127518
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Sport styling with maximum protection
  • Polycarbonate lens absorbs 99.9% UV
  • Nine-base curve lens for wrap and front/side coverage
  • Sleek unisex styling in a comfortable, light polycarbonate frame
  • 3M Catalog# 11384-00000-20, 11385-00000-20, 11386-00000-20, 11388-00000-20, 11542-00000-20, 11543-00000-20, 11544-00000-20, 11743-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Hardcoat
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2010
Lens Tint Light Blue
Frame Color Blue
Model 11543-00000-20
Lens Material High-impact Polycarbonate