Description
- Polycarbonate lens absorbs 99.9% UV
- Nine-base curve lens for wrap and front/side coverage
- Sleek unisex styling in a comfortable, light polycarbonate frame
- 3M Catalog# 11384-00000-20, 11385-00000-20, 11386-00000-20, 11388-00000-20, 11542-00000-20, 11543-00000-20, 11544-00000-20, 11743-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1-2015 |
| Lens Tint | Light Amber |
| Frame Color | Amber |
| Model | 11544-00000-20 |
| Lens Material | High-impact Polycarbonate |