Learn More
3M™ Virtua™ V4 Eyewear
Catalog No. 19161181 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Gray
Catalog No. Lens Tint
19-161-181 Gray
1 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-161-181 Supplier 3M™ Supplier No. 7000029999
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Protection, comfort and style
  • High-impact polycarbonate lens
  • Wraparound lens and unisex styling
  • Soft, dual-injected temples
  • Black temple with gray overmold
  • 3M Catalog# 11674-00000-20, 11673-00000-20, 11672-00000-20, 11671-00000-20, 11670-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Gray
Frame Color Black
Lens Material Polycarbonate