Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Wireless Ergonomic Mouse

Catalog No. 19021854 Shop All 3M Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-021-854 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-021-854 Supplier 3M™ Supplier No. 7000047613

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

AAA Alkaline batteries

Alleviates the pain and discomfort indicative of repetitive stress injuries

Ease-of-Use Commendation from the Arthritis Foundation™

  • Clinically proven
  • Vertical handle design with right and left click buttons keeps wrist in neutral position
  • Third button enables scrolling in some applications
  • Works as a standard optical mouse
  • Wireless, USB compatible plug and play
  • Soft-touch painted surfaces for added comfort
  • For right-handed use
  • Compatible with Windows™ 7 / XP / Vista / 98SE / 2000, and Mac OS 10.6.4 or higher
  • Color: Black
  • Size: Large
  • 3M Catalog# EM550GPL

Warranty and Services

Two year warranty

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Wireless Ergonomic Mouse
Size Large
For Use With (Application) Reduce muscle strain and discomfort associated with Carpal Tunnel Syndrome and repetitive stress injury
Includes AAA alkaline batteries, LED optical sensor
Hand Right
Model EM550GPL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.