Learn More
SOLVENTUM 3M™ Zeta Plus™ BC Series Encapsulated Disposable Filter Capsule 2.63 in, 25 sq cm, EFA, Luer Lock, 4/Case
Supplier:  SOLVENTUM 7010382846
Encompass_Preferred
3M™ Zeta Plus™ Encapsulated Filter Capsules with ZB Grade Media extend depth filtration capacity and enhances the protection of downstream membranes thereby extending filter life. 3M™ Zeta Plus™ ™ Encapsulated Filter Capsules with ZB Grade Media are part of a family of advanced dual zone depth filters designed to provide optimal clarification of bioprocess, biological and pharmaceutical fluids.
Catalog No. NC1759356