Learn More
AdipoGen 3Me-Sulfobutyl-benzindolium
SDP
Catalog No. 502560027
Encompass_Preferred
Chemical. CAS 63149-24-6. Formula C19H23NO3S. MW 345.46. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.
Catalog No. 50-256-0027 Supplier AdipoGen Supplier No. CDXT0061M500
Chemical. CAS 63149-24-6. Formula C19H23NO3S. MW 345.46. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.