Learn More
AdipoGen 2,3,3-Trimethyl-1-(4-sulfobutyl)-indolium, inner salt, 54136-26-4, MFCD08166699, 500 mg
SDP
Supplier:  AdipoGen CDXT0042M500
Encompass_Preferred
MF: C15H21NO3S, MW: 295.4, ≥98%, Appearance: Pink powder. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.
Catalog No. 50-117-7742