Learn More
Cayman Chemical 3methoxy TyramInhydroCLO 100mg
SDP
Catalog No. 502984449
Encompass_Preferred
A natural metabolite of dopamine; a weak agonist of TAAR1 (EC50 = 1.7 µM)
Catalog No. 50-298-4449 Supplier Cayman Chemical Supplier No. 20511100 MG
A natural metabolite of dopamine; a weak agonist of TAAR1 (EC50 = 1.7 µM)