Learn More
Cayman Chemical 3MethoxymethcathInonhydro 10mg
SDP
Catalog No. 502893433
Encompass_Preferred
A positional isomer of methedrone, having the methoxy group at the 3 rather than the 4 position; intended for forensic and research applications
Catalog No. 50-289-3433 Supplier Cayman Chemical Supplier No. 900118710 MG
A positional isomer of methedrone, having the methoxy group at the 3 rather than the 4 position; intended for forensic and research applications