Learn More
Cayman Chemical 3-Methoxymethcathinone (hydrochloride), Purity: ≥98%, CAS: 1435933-70-2, 50 mg
SDP
Catalog No. 502893313
Encompass_Preferred
A positional isomer of methedrone, having the methoxy group at the 3 rather than the 4 position; intended for forensic and research applications
Catalog No. 50-289-3313 Supplier Cayman Chemical Supplier No. 900118750 MG
A positional isomer of methedrone, having the methoxy group at the 3 rather than the 4 position; intended for forensic and research applications