Learn More
Cayman Chemical 3-Methylquinoxaline-2-carboxylic Acid, Purity: ≥98%, CAS: 74003-63-7, 10 mg
SDP
Catalog No. 502934270
Encompass_Preferred
A major metabolite of olaquindox; induces cell cycle arrest at the S phase and is toxic to Chang liver cells in a concentration- and time-dependent manner; has been used as a marker of olaquindox use in livestock applications
Catalog No. 50-293-4270 Supplier Cayman Chemical Supplier No. 2566410 MG
A major metabolite of olaquindox; induces cell cycle arrest at the S phase and is toxic to Chang liver cells in a concentration- and time-dependent manner; has been used as a marker of olaquindox use in livestock applications